Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021322.2 | NP_067297.2 | 7 - 8 | 960 | 79 | |
GenBank mRNA | AJ271892.1 | - | 7 - 8 | 730 | 79 | |
AJ271893.1 | - | 7 - 8 | 730 | 79 | ||
AK035125.1 | - | 6 - 7 | 584 | 79 | ||
AK085245.1 | - | 2 - 3 | 174 | 79 | ||
AK144821.1 | - | 7 - 8 | 765 | 79 | ||
AK152271.1 | - | 7 - 8 | 752 | 79 | ||
AK153161.1 | - | 7 - 8 | 752 | 79 | ||
AK164160.1 | - | 7 - 8 | 81 | |||
BC039272.1 | - | 7 - 8 | 760 | 5027600 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |