Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021334.2 | NP_067309.1 | 2 - 3 | 196 | 89 | |
GenBank mRNA | AF211864.1 | - | 2 - 3 | 196 | 89 | |
AK089167.1 | - | 2 - 3 | 214 | 89 | ||
AK131133.1 | - | 2 - 3 | 202 | 89 | ||
AK155816.1 | - | 2 - 3 | 218 | 89 | ||
AK170432.1 | - | 2 - 3 | 214 | 89 | ||
BC042840.1 | - | 2 - 3 | 197 | 89 | ||
BC057200.1 | - | 2 - 3 | 189 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |