Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021334.2 | NP_067309.1 | 21 - 22 | 2706 | 114 | |
GenBank mRNA | AF211864.1 | - | 21 - 22 | 2706 | 114 | |
AK131133.1 | - | 21 - 22 | 2712 | 114 | ||
AK155816.1 | - | 21 - 22 | 2728 | 114 | ||
AK170432.1 | - | 21 - 22 | 2724 | 114 | ||
BC042840.1 | - | 21 - 22 | 2705 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |