Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021426.4 | NP_067401.3 | 6 - 7 | 693 | 55 | |
GenBank mRNA | AB030182.1 | - | 6 - 7 | 671 | 55 | |
AK005262.1 | - | 6 - 7 | 664 | 55 | ||
AK007632.1 | - | 6 - 7 | 658 | 55 | ||
AK021096.1 | - | 6 - 7 | 688 | 55 | ||
BC061107.1 | - | 6 - 7 | 678 | 30295742 | 55 | |
BC089330.1 | - | 6 - 7 | 645 | 30358178 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |