Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021877.3 | NP_068677.2 | 3 - 4 | 1306 | 67 | |
GenBank mRNA | AB221597.1 | - | 1 - 2 | 612 | 67 | |
AY312855.1 | - | 2 - 3 | 989 | 67 | ||
AY547390.1 | - | 2 - 3 | 989 | 67 | ||
AY547391.1 | - | 2 - 3 | 989 | 67 | ||
BC049182.1 | - | 3 - 4 | 1306 | 6468299 | 67 | |
Z32675.1 | - | 2 - 3 | 989 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |