Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021877.3 | NP_068677.2 | 6 - 7 | 2431 | 81 | |
GenBank mRNA | AB221596.1 | - | 1 - 2 | 116 | 81 | |
AB221597.1 | - | 4 - 5 | 1734 | 81 | ||
AY312855.1 | - | 5 - 6 | 2114 | 81 | ||
AY547390.1 | - | 5 - 6 | 2111 | 81 | ||
AY547391.1 | - | 5 - 6 | 2111 | 81 | ||
BC049182.1 | - | 6 - 7 | 2431 | 6468299 | 81 | |
Z32675.1 | - | 5 - 6 | 2114 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |