Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021879.2 | NP_068679.1 | 19 - 20 | 2196 | 86 | |
GenBank mRNA | AK051897.1 | - | 18 - 19 | 2037 | 86 | |
AK087494.1 | - | 20 - 21 | 2456 | 86 | ||
BC119220.1 | - | 19 - 20 | 2166 | 8733969 | 86 | |
BC120549.1 | - | 19 - 20 | 2166 | 40129472 | 86 | |
M97900.1 | - | 19 - 20 | 2196 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |