Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199118.1 | NP_001186047.1 | 2 - 3 | 443 | 92 | |
NM_022883.3 | NP_075021.1 | 2 - 3 | 520 | 92 | ||
GenBank mRNA | AF286724.1 | - | 2 - 3 | 457 | 92 | |
AK033647.1 | - | 2 - 3 | 441 | 92 | ||
AK049363.1 | - | 2 - 3 | 436 | 92 | ||
AK163833.1 | - | 2 - 3 | 435 | 92 | ||
AK220228.1 | - | 2 - 3 | 520 | 92 | ||
BB659464.1 | - | 2 - 3 | 520 | 92 | ||
BC026729.1 | - | 2 - 3 | 427 | 92 | ||
BC117882.1 | - | 1 - 2 | 384 | 40096038 | 92 | |
BC117883.1 | - | 1 - 2 | 384 | 40096041 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |