Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199118.1 | NP_001186047.1 | 12 - 13 | 1863 | 65 | |
NM_022883.3 | NP_075021.1 | 12 - 13 | 1940 | 65 | ||
GenBank mRNA | AF286724.1 | - | 12 - 13 | 1877 | 65 | |
AK033647.1 | - | 12 - 13 | 1891 | 65 | ||
AK049363.1 | - | 12 - 13 | 1856 | 65 | ||
AK163833.1 | - | 12 - 13 | 1855 | 65 | ||
AK220228.1 | - | 12 - 13 | 1940 | 65 | ||
BC026729.1 | - | 12 - 13 | 1847 | 65 | ||
BC117882.1 | - | 11 - 12 | 1804 | 40096038 | 65 | |
BC117883.1 | - | 11 - 12 | 1804 | 40096041 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |