Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085383.1 | NP_001078852.1 | 7 - 8 | 696 | 113 | |
NM_023628.2 | NP_076117.2 | 7 - 8 | 675 | 113 | ||
GenBank mRNA | AJ401160.1 | - | 7 - 8 | 629 | 113 | |
AK003395.1 | - | 7 - 8 | 696 | 113 | ||
AK010128.1 | - | 7 - 8 | 676 | 113 | ||
BC062140.1 | - | 7 - 8 | 686 | 30290956 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |