Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800012673.mm.1 |
NM_023709.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012673.mm.1 |
NM_023709.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012673.mm.1 |
TC0800001545.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012676.mm.1 |
NM_023709.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012676.mm.1 |
NM_023709.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012676.mm.1 |
TC0800001545.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012672.mm.1 |
NM_023709.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012672.mm.1 |
NM_023709.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012672.mm.1 |
TC0800001545.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012674.mm.1 |
NM_023709.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012674.mm.1 |
NM_023709.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012674.mm.1 |
TC0800001545.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012675.mm.1 |
NM_023709.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012675.mm.1 |
NM_023709.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012675.mm.1 |
TC0800001545.mm.1 |
MTA TC |