Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159965.1 | NP_001153437.1 | 13 - 14 | 1430 | 71 | |
NM_001159966.1 | NP_001153438.1 | 13 - 14 | 1328 | 71 | ||
NM_001159967.1 | NP_001153439.1 | 13 - 14 | 1430 | 71 | ||
NM_001159968.1 | NP_001153440.1 | 12 - 13 | 1325 | 71 | ||
NM_023884.4 | NP_076373.3 | 13 - 14 | 1370 | 71 | ||
NR_027640.1 | 14 - 15 | 1535 | 71 | |||
GenBank mRNA | AF312924.1 | - | 12 - 13 | 1278 | 71 | |
AK018622.1 | - | 13 - 14 | 1372 | 71 | ||
AK019543.1 | - | 12 - 13 | 1328 | 71 | ||
AK029049.1 | - | 13 - 14 | 1371 | 71 | ||
AK029993.1 | - | 14 - 15 | 1508 | 71 | ||
AK033549.1 | - | 13 - 14 | 1326 | 71 | ||
AK036803.1 | - | 12 - 13 | 1302 | 71 | ||
AK161080.1 | - | 13 - 14 | 1397 | 71 | ||
AK165002.1 | - | 13 - 14 | 1322 | 71 | ||
AK169368.1 | - | 13 - 14 | 1348 | 71 | ||
BC043132.1 | - | 13 - 14 | 1335 | 6811441 | 71 | |
BC052663.1 | - | 13 - 14 | 1347 | 30051209 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |