Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025808.3 | NP_080084.2 | 16 - 17 | 1995 | 66 | |
GenBank mRNA | AK004561.2 | - | 16 - 17 | 1995 | 66 | |
AK005037.1 | - | 16 - 17 | 1994 | 66 | ||
AK083411.1 | - | 16 - 17 | 1999 | 66 | ||
AK088938.1 | - | 7 - 8 | 927 | 66 | ||
AK132817.1 | - | 16 - 17 | 1994 | 66 | ||
AK208308.1 | - | 1 - 2 | 44 | 66 | ||
AK219434.1 | - | 1 - 2 | 44 | 66 | ||
BC034400.1 | - | 16 - 17 | 1989 | 2651456 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |