Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026384.3 | NP_080660.1 | 1 - 2 | 320 | 81 | |
GenBank mRNA | AF384160.1 | - | 1 - 2 | 178 | 81 | |
AK002443.1 | - | 1 - 2 | 325 | 81 | ||
AK016660.1 | - | 1 - 2 | 268 | 81 | ||
BC043447.1 | - | 1 - 2 | 320 | 5125951 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |