Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053191.2 | NP_444421.2 | 4 - 5 | 696 | 96 | |
GenBank mRNA | AF329196.1 | - | 4 - 5 | 616 | 96 | |
AK030960.1 | - | 4 - 5 | 698 | 96 | ||
AK040961.1 | - | 4 - 5 | 695 | 96 | ||
AK165584.1 | - | 3 - 4 | 372 | 96 | ||
BC116705.1 | - | 4 - 5 | 592 | 40102877 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |