Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938926 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938931 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938929 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938928 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938930 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118784 |
BC055719.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118784 |
X83587.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118784 |
NM_054048.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118784 |
AK052295.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118784 |
AK205817.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
483921 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824861 |
AK054426.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824861 |
AY355342.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824861 |
BC066795.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824861 |
NM_172481.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
124395 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114831 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118784 |
BC055719.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118784 |
X83587.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118784 |
NM_054048.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118784 |
AK052295.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118784 |
AK205817.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306689 |
BC055719.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306689 |
X83587.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306689 |
NM_054048.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306689 |
AK052295.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P306689 |
AK205817.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938913 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938929 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938923 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938884 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938910 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938919 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938926 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938890 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938907 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938942 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938888 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938909 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938924 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938945 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938946 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938933 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938931 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938883 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938906 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938938 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938920 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938911 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938947 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938940 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938892 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938912 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938935 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938921 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938915 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938932 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938887 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938917 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938916 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938895 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938899 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938898 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938897 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938904 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938937 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938908 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938927 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938903 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938885 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31938902 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114831 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03546009 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P306689 |
BC055719.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306689 |
X83587.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306689 |
NM_054048.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306689 |
AK052295.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P306689 |
AK205817.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105984 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114831 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14419381 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14190933 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14190930 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14190927 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938929 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938928 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938930 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938931 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31938926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938895 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938937 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31939054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31938917 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938908 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14301706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938902 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938892 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14273090 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938935 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938970 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938923 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938974 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938963 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14412832 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938888 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938932 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938947 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14314893 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14400730 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938942 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938907 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938915 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938897 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14393124 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938933 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938949 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938870 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938861 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938966 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938873 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938862 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938955 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938903 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938951 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938953 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938911 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938866 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14190922 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938912 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938885 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14451021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14436666 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938883 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14190924 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14300753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938868 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938956 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938871 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938917 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938946 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938863 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938958 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14257113 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938906 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14273087 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14273091 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14301707 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938937 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14301689 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14442497 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14436667 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938887 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938931 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938880 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938957 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938864 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938929 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14393137 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938973 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31938890 |
- |
- |