Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rogdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133185.2 | NP_573448.2 | 6 - 7 | 473 | 72 | |
GenBank mRNA | AK002523.1 | - | 6 - 7 | 457 | 72 | |
AK046477.1 | - | 6 - 7 | 434 | 72 | ||
AK170144.1 | - | 6 - 7 | 438 | 72 | ||
AY036117.1 | - | 6 - 7 | 441 | 72 | ||
BC006914.1 | - | 6 - 7 | 516 | 3601096 | 72 | |
BU604779.1 | - | 6 - 7 | 490 | 72 | ||
CD363552.1 | - | 1 - 2 | 39 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |