Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122604.1 | NP_001116076.1 | 9 - 10 | 893 | 124 | |
NM_001122605.1 | NP_001116077.1 | 9 - 10 | 890 | 124 | ||
NM_133188.2 | NP_573451.2 | 9 - 10 | 890 | 124 | ||
GenBank mRNA | AF225910.1 | - | 9 - 10 | 124 | ||
AK148023.1 | - | 9 - 10 | 768 | 124 | ||
AK152314.1 | - | 9 - 10 | 860 | 124 | ||
AK152345.1 | - | 9 - 10 | 860 | 124 | ||
BC049355.1 | - | 9 - 10 | 810 | 6337388 | 124 | |
BC058775.1 | - | 9 - 10 | 804 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |