Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290398.1 | NP_001277327.1 | 1 - 2 | 249 | 79 | |
NM_001290399.1 | NP_001277328.1 | 1 - 2 | 249 | 79 | ||
NM_133196.6 | NP_573459.1 | 1 - 2 | 249 | 79 | ||
GenBank mRNA | AF317552.1 | - | 1 - 2 | 217 | 79 | |
AK032817.1 | - | 1 - 2 | 231 | 79 | ||
AK049971.1 | - | 1 - 2 | 239 | 79 | ||
AK088260.1 | - | 1 - 2 | 77 | 79 | ||
BC036719.1 | - | 1 - 2 | 221 | 5322335 | 79 | |
CJ183629.1 | - | 1 - 2 | 249 | 79 | ||
EU616680.1 | - | 1 - 2 | 217 | 79 | ||
EU616681.1 | - | 1 - 2 | 217 | 79 | ||
EU616682.1 | - | 1 - 2 | 217 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |