Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290398.1 | NP_001277327.1 | 5 - 6 | 754 | 62 | |
NM_001290399.1 | NP_001277328.1 | 5 - 6 | 754 | 62 | ||
NM_133196.6 | NP_573459.1 | 5 - 6 | 754 | 62 | ||
GenBank mRNA | AF317552.1 | - | 5 - 6 | 722 | 62 | |
AK032817.1 | - | 5 - 6 | 736 | 62 | ||
AK049971.1 | - | 5 - 6 | 745 | 62 | ||
AK088260.1 | - | 5 - 6 | 582 | 62 | ||
AK147438.1 | - | 4 - 5 | 520 | 62 | ||
BC036719.1 | - | 5 - 6 | 726 | 5322335 | 62 | |
EU616680.1 | - | 5 - 6 | 722 | 62 | ||
EU616681.1 | - | 5 - 6 | 722 | 62 | ||
EU616682.1 | - | 5 - 6 | 722 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |