Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133198.2 | NP_573461.2 | 5 - 6 | 754 | 134 | |
GenBank mRNA | AF288783.1 | - | 5 - 6 | 696 | 134 | |
AK140321.1 | - | 5 - 6 | 701 | 134 | ||
AK145989.1 | - | 5 - 6 | 755 | 134 | ||
AK149491.1 | - | 929 | 134 | |||
AK167337.1 | - | 5 - 6 | 701 | 134 | ||
BC013636.1 | - | 5 - 6 | 688 | 4196237 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |