Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133198.2 | NP_573461.2 | 10 - 11 | 1337 | 84 | |
GenBank mRNA | AF288783.1 | - | 10 - 11 | 1279 | 84 | |
AK140321.1 | - | 10 - 11 | 1284 | 84 | ||
AK145989.1 | - | 10 - 11 | 1338 | 84 | ||
AK149491.1 | - | 1512 | 84 | |||
AK167337.1 | - | 10 - 11 | 1284 | 84 | ||
BC013636.1 | - | 10 - 11 | 1271 | 4196237 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |