Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285920.1 | NP_001272849.1 | 13 - 14 | 1772 | 73 | |
NM_001285921.1 | NP_001272850.1 | 13 - 14 | 1755 | 73 | ||
NM_001285922.1 | NP_001272851.1 | 12 - 13 | 1686 | 73 | ||
NM_001285923.1 | NP_001272852.1 | 12 - 13 | 1686 | 73 | ||
NM_133201.3 | NP_573464.2 | 12 - 13 | 1689 | 73 | ||
GenBank mRNA | AF384100.1 | - | 13 - 14 | 1569 | 73 | |
AK042080.1 | - | 12 - 13 | 1470 | 73 | ||
AK049583.1 | - | 12 - 13 | 1479 | 73 | ||
AK052689.1 | - | 12 - 13 | 1470 | 73 | ||
AK122220.1 | - | 11 - 12 | 2664 | 73 | ||
AK146801.1 | - | 13 - 14 | 1532 | 73 | ||
AK147863.1 | - | 12 - 13 | 1485 | 73 | ||
AK169373.1 | - | 12 - 13 | 1474 | 73 | ||
AY028170.1 | - | 1450 | 73 | |||
AY123975.1 | - | 12 - 13 | 1636 | 73 | ||
BC046503.1 | - | 12 - 13 | 1467 | 4191179 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |