Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK077593.1 | - | 7 - 8 | 1321 | 65 | |
AK141558.1 | - | 7 - 8 | 1352 | 65 | ||
AK165931.1 | - | 7 - 8 | 1318 | 65 | ||
BC050195.1 | - | 7 - 8 | 1297 | 30010078 | 65 | |
U70859.1 | - | 8 - 9 | 1455 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |