Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301250.1 | NP_001288179.1 | 2 - 3 | 258 | 77 | |
NM_007602.4 | NP_031628.1 | 2 - 3 | 422 | 77 | ||
GenBank mRNA | AK155179.1 | - | 2 - 3 | 400 | 77 | |
AK164293.1 | - | 2 - 3 | 332 | 77 | ||
AK170034.1 | - | 2 - 3 | 359 | 77 | ||
BC014767.1 | - | 2 - 3 | 250 | 4458488 | 77 | |
U85020.1 | - | 2 - 3 | 341 | 77 | ||
Y10656.1 | - | 2 - 3 | 354 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |