Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008036.2 | NP_032062.1 | 2 - 3 | 1055 | 66 | |
GenBank mRNA | BC132064.1 | - | 2 - 3 | 610 | 40130341 | 66 |
BC137994.1 | - | 2 - 3 | 610 | 8861013 | 66 | |
BC145082.1 | - | 2 - 3 | 610 | 66 | ||
CK795975.1 | - | 1 - 2 | 156 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |