Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271497.1 | NP_001258426.1 | 3 - 4 | 459 | 96 | |
NM_008358.2 | NP_032384.1 | 3 - 4 | 522 | 96 | ||
NM_133836.2 | NP_598597.1 | 3 - 4 | 575 | 96 | ||
GenBank mRNA | AY221616.1 | - | 3 - 4 | 200 | 96 | |
BC022705.1 | - | 3 - 4 | 332 | 96 | ||
BC132231.1 | - | 2 - 3 | 122 | 96 | ||
BC132233.1 | - | 2 - 3 | 122 | 96 | ||
BC145348.1 | - | 2 - 3 | 122 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |