Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113209.2 | NP_001106680.1 | 4 - 5 | 1709 | 80 | |
NM_001113210.2 | NP_001106681.1 | 4 - 5 | 1709 | 80 | ||
NM_001286127.1 | NP_001273056.1 | 4 - 5 | 1709 | 80 | ||
NM_001286131.1 | NP_001273060.1 | 4 - 5 | 1709 | 80 | ||
NM_008687.6 | NP_032713.3 | 4 - 5 | 1709 | 80 | ||
GenBank mRNA | AK034261.1 | - | 4 - 5 | 1241 | 80 | |
AK083167.1 | - | 4 - 5 | 1054 | 80 | ||
AK145267.1 | - | 3 - 4 | 415 | 80 | ||
AK146480.1 | - | 4 - 5 | 1226 | 80 | ||
AK148573.1 | - | 4 - 5 | 1697 | 80 | ||
AY035852.1 | - | 4 - 5 | 714 | 80 | ||
AY035853.1 | - | 4 - 5 | 714 | 80 | ||
BC014290.1 | - | 4 - 5 | 746 | 4038233 | 80 | |
BC023366.1 | - | 4 - 5 | 846 | 80 | ||
BC062908.1 | - | 4 - 5 | 1031 | 6815775 | 80 | |
BE862566.1 | - | 3 - 4 | 360 | 80 | ||
U57634.1 | - | 4 - 5 | 915 | 80 | ||
Y07685.1 | - | 4 - 5 | 1641 | 80 | ||
Y07686.1 | - | 4 - 5 | 1221 | 80 | ||
Y07687.1 | - | 4 - 5 | 1702 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |