Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008718.2 | NP_032744.1 | 2 - 3 | 316 | 70 | |
GenBank mRNA | AK158830.1 | - | 2 - 3 | 316 | 70 | |
BC132113.1 | - | 1 - 2 | 151 | 40130390 | 70 | |
BC137863.1 | - | 1 - 2 | 151 | 8860882 | 70 | |
BY279348.1 | - | 2 - 3 | 316 | 70 | ||
U77967.1 | - | 1 - 2 | 184 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |