Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008718.2 | NP_032744.1 | 6 - 7 | 880 | 121 | |
GenBank mRNA | AK158830.1 | - | 6 - 7 | 960 | 121 | |
BC132113.1 | - | 5 - 6 | 715 | 40130390 | 121 | |
BC137863.1 | - | 5 - 6 | 715 | 8860882 | 121 | |
U77967.1 | - | 5 - 6 | 748 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |