Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008744.2 | NP_032770.2 | 4 - 5 | 1638 | 136 | |
GenBank mRNA | AF128865.1 | - | 3 - 4 | 1373 | 136 | |
BC141294.1 | - | 3 - 4 | 1407 | 9055906 | 136 | |
BE652257.1 | - | 3 - 4 | 136 | |||
U65418.1 | - | 3 - 4 | 1354 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |