Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008770.3 | NP_032796.1 | 1 - 2 | 448 | 68 | |
GenBank mRNA | AF124426.1 | - | 1 - 2 | 218 | 68 | |
AK005088.1 | - | 1 - 2 | 423 | 68 | ||
AK005171.1 | - | 1 - 2 | 423 | 68 | ||
AK161698.1 | - | 1 - 2 | 423 | 68 | ||
BC021659.1 | - | 1 - 2 | 400 | 4190031 | 68 | |
U19582.1 | - | 1 - 2 | 377 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |