Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008779.2 | NP_032805.2 | 15 - 16 | 2176 | 79 | |
GenBank mRNA | AK140747.1 | - | 6 - 7 | 765 | 79 | |
L01991.1 | - | 2285 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |