Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163688.1 | NP_001157160.1 | 15 - 16 | 1963 | 72 | |
NM_008980.2 | NP_033006.2 | 16 - 17 | 2071 | 72 | ||
GenBank mRNA | AF332081.1 | - | 14 - 15 | 1676 | 72 | |
AF332082.1 | - | 14 - 15 | 1676 | 72 | ||
AJ431367.1 | - | 14 - 15 | 1676 | 72 | ||
AK031690.1 | - | 15 - 16 | 1962 | 72 | ||
AK165673.1 | - | 15 - 16 | 1961 | 72 | ||
BC050930.1 | - | 15 - 16 | 1945 | 6811172 | 72 | |
BC110683.1 | - | 15 - 16 | 1929 | 3965311 | 72 | |
M36033.1 | - | 16 - 17 | 1809 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |