Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009020.3 | NP_033046.1 | 1 - 2 | 123 | 82 | |
GenBank mRNA | AK031014.1 | - | 1 - 2 | 133 | 82 | |
AK040375.1 | - | 1 - 2 | 124 | 82 | ||
AK089075.1 | - | 1 - 2 | 155 | 82 | ||
BC120548.1 | - | 1 - 2 | 80 | 40129471 | 82 | |
BC125473.1 | - | 1 - 2 | 80 | 40129988 | 82 | |
M64796.1 | - | 1 - 2 | 67 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |