Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009056.2 | NP_033082.1 | 3 - 4 | 312 | 95 | |
NM_027787.1 | NP_082063.1 | 3 - 4 | 312 | 95 | ||
GenBank mRNA | AK044439.1 | - | 3 - 4 | 312 | 95 | |
AK086559.1 | - | 3 - 4 | 244 | 95 | ||
BC004654.1 | - | 3 - 4 | 329 | 3497910 | 95 | |
X76089.1 | - | 3 - 4 | 267 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |