Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009063.3 | NP_033089.2 | 3 - 4 | 300 | 95 | |
GenBank mRNA | AK004165.1 | - | 3 - 4 | 301 | 95 | |
AK028765.1 | - | 302 | 95 | |||
AK030300.1 | - | 96 | ||||
AK044096.1 | - | 3 - 4 | 301 | 95 | ||
AK054098.1 | - | 3 - 4 | 301 | 95 | ||
BC037683.1 | - | 3 - 4 | 287 | 3962083 | 95 | |
U67188.1 | - | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |