Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037726.1 | NP_001032815.1 | 3 - 4 | 468 | 106 | |
NM_009952.2 | NP_034082.1 | 5 - 6 | 632 | 106 | ||
NM_133828.2 | NP_598589.2 | 4 - 5 | 590 | 106 | ||
GenBank mRNA | AF448507.1 | - | 5 - 6 | 508 | 106 | |
AF448508.1 | - | 1160 | 106 | |||
AK042595.1 | - | 4 - 5 | 563 | 106 | ||
AK044992.1 | - | 4 - 5 | 405 | 106 | ||
AK044993.1 | - | 4 - 5 | 405 | 106 | ||
AK079223.1 | - | 4 - 5 | 586 | 106 | ||
BC021649.1 | - | 4 - 5 | 523 | 4017170 | 106 | |
M95106.1 | - | 5 - 6 | 542 | 106 | ||
U46027.1 | - | 3 - 4 | 338 | 106 | ||
X92497.1 | - | 3 - 4 | 445 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |