Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010819.4 | NP_034949.3 | 3 - 4 | 453 | 98 | |
GenBank mRNA | AF061272.1 | - | 3 - 4 | 385 | 98 | |
AK089500.1 | - | 3 - 4 | 444 | 98 | ||
AK171409.1 | - | 3 - 4 | 391 | 98 | ||
BC099471.1 | - | 3 - 4 | 363 | 30845162 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |