Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 30 - 31 | 4492 | 64 | |
GenBank mRNA | AF093576.1 | - | 30 - 31 | 4400 | 64 | |
AK161449.1 | - | 1 - 2 | 133 | 64 | ||
BC150747.1 | - | 30 - 31 | 4357 | 9087658 | 64 | |
U87455.1 | - | 30 - 31 | 4472 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |