Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011851.4 | NP_035981.1 | 2 - 3 | 646 | 122 | |
GenBank mRNA | AK028723.1 | - | 2 - 3 | 616 | 122 | |
AK029979.1 | - | 2 - 3 | 616 | 122 | ||
AK154614.1 | - | 2 - 3 | 623 | 122 | ||
BC119268.1 | - | 2 - 3 | 590 | 8734017 | 122 | |
BC119270.1 | - | 2 - 3 | 590 | 8734019 | 122 | |
L12059.1 | - | 2 - 3 | 569 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |