Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080118.1 | NP_001073587.1 | 4 - 5 | 478 | 125 | |
NM_013634.2 | NP_038662.2 | 4 - 5 | 343 | 125 | ||
NM_134027.2 | NP_598788.2 | 4 - 5 | 478 | 125 | ||
GenBank mRNA | AF000294.1 | - | 4 - 5 | 343 | 125 | |
AF332073.1 | - | 3 - 4 | 223 | 125 | ||
AF332074.1 | - | 3 - 4 | 223 | 125 | ||
AK049203.1 | - | 4 - 5 | 469 | 125 | ||
AK054437.1 | - | 4 - 5 | 479 | 125 | ||
AK147199.1 | - | 4 - 5 | 455 | 125 | ||
BC021440.1 | - | 4 - 5 | 449 | 125 | ||
BC079636.1 | - | 4 - 5 | 448 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |