Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019587.2 | NP_062533.2 | 16 - 17 | 2930 | 67 | |
GenBank mRNA | AK048728.1 | - | 2 - 3 | 228 | 67 | |
AK138168.1 | - | 14 - 15 | 1757 | 67 | ||
AK138631.1 | - | 15 - 16 | 1853 | 67 | ||
AK140240.1 | - | 16 - 17 | 2934 | 67 | ||
AK170004.1 | - | 2 - 3 | 276 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |