Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019587.2 | NP_062533.2 | 22 - 23 | 3953 | 94 | |
GenBank mRNA | AK138168.1 | - | 18 - 19 | 2958 | 94 | |
AK140240.1 | - | 20 - 21 | 4135 | 94 | ||
AK170004.1 | - | 9 - 10 | 1203 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |