Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019696.2 | NP_062670.2 | 10 - 11 | 1514 | 63 | |
GenBank mRNA | AF077738.1 | - | 10 - 11 | 1459 | 63 | |
AK132611.1 | - | 6 - 7 | 760 | 63 | ||
BC003713.1 | - | 10 - 11 | 1447 | 3593211 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |