Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167799.1 | NP_001161271.1 | 9 - 10 | 952 | 83 | |
NM_019829.4 | NP_062803.4 | 8 - 9 | 1251 | 83 | ||
GenBank mRNA | AF232709.1 | - | 8 - 9 | 619 | 83 | |
AK166087.1 | - | 8 - 9 | 1254 | 83 | ||
BC004849.1 | - | 9 - 10 | 885 | 2655845 | 83 | |
BC021883.1 | - | 8 - 9 | 1234 | 3487476 | 83 | |
DV649852.1 | - | 3 - 4 | 215 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |