Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022654.1 | NP_073145.1 | 6 - 7 | 1397 | 79 | |
GenBank mRNA | AF274973.1 | - | 6 - 7 | 1397 | 79 | |
AK004702.1 | - | 7 - 8 | 1340 | 79 | ||
AK156863.1 | - | 8 - 9 | 1385 | 79 | ||
BC032202.1 | - | 6 - 7 | 1358 | 79 | ||
BC056987.1 | - | 6 - 7 | 1748 | 79 | ||
BC089373.1 | - | 6 - 7 | 1369 | 79 | ||
BC099686.1 | - | 6 - 7 | 1735 | 79 | ||
BC145314.1 | - | 6 - 7 | 1401 | 79 | ||
BC145315.1 | - | 6 - 7 | 1362 | 79 | ||
BC145857.1 | - | 6 - 7 | 1362 | 40131160 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |