Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022654.1 | NP_073145.1 | 10 - 11 | 2008 | 76 | |
GenBank mRNA | AF274973.1 | - | 10 - 11 | 2008 | 76 | |
AK004702.1 | - | 10 - 11 | 2028 | 76 | ||
AK156863.1 | - | 12 - 13 | 1996 | 76 | ||
BC032202.1 | - | 10 - 11 | 1885 | 76 | ||
BC056987.1 | - | 10 - 11 | 2359 | 76 | ||
BC089373.1 | - | 10 - 11 | 1980 | 76 | ||
BC099686.1 | - | 10 - 11 | 2346 | 76 | ||
BC145314.1 | - | 10 - 11 | 2012 | 76 | ||
BC145315.1 | - | 9 - 10 | 2050 | 76 | ||
BC145857.1 | - | 10 - 11 | 1973 | 40131160 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |