Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023055.2 | NP_075542.2 | 5 - 6 | 946 | 71 | |
NM_023449.3 | NP_075938.2 | 4 - 5 | 516 | 71 | ||
GenBank mRNA | AB026489.1 | - | 5 - 6 | 919 | 71 | |
AK002557.1 | - | 4 - 5 | 517 | 71 | ||
AK039491.1 | - | 5 - 6 | 934 | 71 | ||
AK075800.1 | - | 5 - 6 | 949 | 71 | ||
AK075813.1 | - | 5 - 6 | 947 | 71 | ||
AK084801.1 | - | 5 - 6 | 947 | 71 | ||
AK169007.1 | - | 5 - 6 | 937 | 71 | ||
AK170057.1 | - | 5 - 6 | 932 | 71 | ||
AK179335.1 | - | 2 - 3 | 90 | 71 | ||
BC065778.1 | - | 4 - 5 | 487 | 3467172 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |