Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023630.2 | NP_076119.2 | 2 - 3 | 180 | 76 | |
GenBank mRNA | AF250835.1 | - | 2 - 3 | 148 | 76 | |
AK005881.1 | - | 2 - 3 | 155 | 76 | ||
BC050756.1 | - | 2 - 3 | 163 | 6741860 | 76 | |
BY092414.1 | - | 2 - 3 | 180 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |